CAS RN 4682-36-4
CAS RN 4682-36-4
IUPAC Name:
2-hydroxypropane-1,2,3-tricarboxylic acid; dimethyl({2-[(2-methylphenyl)(phenyl)methoxy]ethyl})amine
Synonyms:
flexon norflex orphenadrine citrate banflex X-otag orphenadrine dihydrogen citrate
Molecular Weight (g/mol):
461.51
Molecular Formula:
C24H31NO8
InChi Key:
MMMNTDFSPSQXJP-UHFFFAOYNA-N
SMILES:
OC(=O)CC(O)(CC(O)=O)C(O)=O.CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1C