CAS RN 469-77-2
IUPAC Name:
2-(1-carboxyundecyl)-5-oxooxolane-2-carboxylic acid
Synonyms:
spiculisporic acid
Molecular Weight (g/mol):
328.41
Molecular Formula:
C17H28O6
InChi Key:
TUXHHVJPGQUPCF-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCC(C(O)=O)C1(CCC(=O)O1)C(O)=O
1
–
1
of
1
results
1
4,5-Dicarboxy-gamma-pentadecanolactone 98.0+%, TCI America™
CAS: 469-77-2 Molecular Formula: C17H28O6 Molecular Weight (g/mol): 328.405 MDL Number: MFCD00059059 InChI Key: TUXHHVJPGQUPCF-SUMWQHHRSA-N Synonym: Spiculisporic Acid PubChem CID: 95228527 IUPAC Name: (2R)-2-[(1R)-1-carboxyundecyl]-5-oxooxolane-2-carboxylic acid SMILES: CCCCCCCCCCC(C(=O)O)C1(CCC(=O)O1)C(=O)O