accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4692-98-2

IUPAC Name: 6-bromo-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Synonyms: 6-bromo-1H-3,1-benzoxazine-2,4-dione
Molecular Weight (g/mol): 242.03
Molecular Formula: C8H4BrNO3
InChi Key: DXSMYDSFWCOSFM-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC(=O)OC(=O)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results