accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOH3COClClCl(Z)

CAS RN 470-90-6

IUPAC Name: (1Z)-2-chloro-1-(2,4-dichlorophenyl)ethenyl diethyl phosphate
Synonyms: chlorfenvinphos
Molecular Weight (g/mol): 359.56
Molecular Formula: C12H14Cl3O4P
InChi Key: FSAVDKDHPDSCTO-WQLSENKSSA-N
SMILES: CCOP(=O)(OCC)O\C(=C/Cl)C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results