accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3CSSiCH3CH3H3C

CAS RN 4712-51-0

IUPAC Name: trimethyl({[(trimethylsilyl)methyl]sulfanyl}methyl)silane
Synonyms: trimethyl({[(trimethylsilyl)methyl]sulfanyl}methyl)silane
Molecular Weight (g/mol): 206.49
Molecular Formula: C8H22SSi2
InChi Key: GISUCMFTNKRCPF-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CSC[Si](C)(C)C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)