accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4721-16-8

IUPAC Name: heptylphosphonic acid
Synonyms: heptylphosphonic acid
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H17O3P
InChi Key: VAJFLSRDMGNZJY-UHFFFAOYSA-N
SMILES: CCCCCCCP(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results