accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 47230-38-6

IUPAC Name: 4,4'-diethyl [1,1'-biphenyl]-4,4'-dicarboxylate
Synonyms: 4,4'-diethyl [1,1'-biphenyl]-4,4'-dicarboxylate
Molecular Weight (g/mol): 298.34
Molecular Formula: C18H18O4
InChi Key: SYTZNHBXNLYWAK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)OCC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)