Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl biphenyl-4,4'-dicarboxylate, 98%, Thermo Scientific™

Catalog No. AAA1455203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1455203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1455203 Supplier Thermo Scientific Chemicals Supplier No. A1455203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47230-38-6
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.34
MDL Number MFCD00017270
InChI Key SYTZNHBXNLYWAK-UHFFFAOYSA-N
Synonym diethyl 4,4'-biphenyldicarboxylate, diethyl biphenyl-4,4'-dicarboxylate, diethyl 1,1'-biphenyl-4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonylphenyl benzoate, diethyl-4,4'-diphenyldicarboxylate, diethyl biphenyl 4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonyl phenyl benzoate, 4,4'-biphenyldicarboxylic acid diethyl ester, 4,4'-diethyl 1,1'-biphenyl-4,4'-dicarboxylate, acmc-1apwf
PubChem CID 261553
IUPAC Name 4,4'-diethyl [1,1'-biphenyl]-4,4'-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 112°C to 114°C
Quantity 1 g
Formula Weight 298.34
Percent Purity 98%
Chemical Name or Material Diethyl biphenyl-4,4'-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.