Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 4,4'-Biphenyldicarboxylate 98.0+%, TCI America™
SDP

Catalog No. B08315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08315G 5 g
B08311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08315G Supplier TCI America Supplier No. B08315G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 47230-38-6
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.34
MDL Number MFCD00017270
InChI Key SYTZNHBXNLYWAK-UHFFFAOYSA-N
Synonym diethyl 4,4'-biphenyldicarboxylate, diethyl biphenyl-4,4'-dicarboxylate, diethyl 1,1'-biphenyl-4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonylphenyl benzoate, diethyl-4,4'-diphenyldicarboxylate, diethyl biphenyl 4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonyl phenyl benzoate, 4,4'-biphenyldicarboxylic acid diethyl ester, 4,4'-diethyl 1,1'-biphenyl-4,4'-dicarboxylate, acmc-1apwf
PubChem CID 261553
IUPAC Name 4,4'-diethyl [1,1'-biphenyl]-4,4'-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
Formula Weight 298.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 4,4′-Biphenyldicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.