accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2FFFFFF

CAS RN 47250-53-3

IUPAC Name: 3-[2-(3-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
Synonyms: 3-[2-(3-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
Molecular Weight (g/mol): 334.27
Molecular Formula: C15H12F6N2
InChi Key: UVUCUHVQYAPMEU-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C(C1=CC(N)=CC=C1)(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results