CAS RN 473728-67-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ CXCR2 antagonist, Cpd 19
The CXCR2 antagonist, Cpd 19, also referenced under CAS 473728-67-5, controls the biological activity of CXCR2.