accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4744-10-9

IUPAC Name: 1,1-dimethoxypropane
Synonyms: 1,1-dimethoxypropane
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: UIOXNNAWANDJCZ-UHFFFAOYSA-N
SMILES: CCC(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results