Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionaldehyde Dimethyl Acetal 98.0+%, TCI America™
SDP

Catalog No. P078225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P078225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P078225ML Supplier TCI America Supplier No. P078225ML

Chemical Identifiers

CAS 4744-10-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00059307
InChI Key UIOXNNAWANDJCZ-UHFFFAOYSA-N
Synonym 1,1-Dimethoxypropane
PubChem CID 20859
IUPAC Name 1,1-dimethoxypropane
SMILES CCC(OC)OC

Specifications

Color Yellow
Boiling Point 89°C
Quantity 25 mL
UN Number 1993
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propionaldehyde Dimethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.