accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4747-07-3

IUPAC Name: 1-methoxyhexane
Synonyms: hexane, 1-methoxy-
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: ICBJCVRQDSQPGI-UHFFFAOYSA-N
SMILES: CCCCCCOC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results