Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Methyl Ether 98.0+%, TCI America™
SDP

Catalog No. H093025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H093025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H093025ML Supplier TCI America Supplier No. H093025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4747-07-3
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00015350
InChI Key ICBJCVRQDSQPGI-UHFFFAOYSA-N
Synonym hexyl methyl ether, hexane, 1-methoxy, methyl hexyl ether, ether, hexyl methyl, methyl-n-hexyl ether, n-hexyl methyl ether, unii-r99k6anq7t, methoxyhexane, r99k6anq7t, 2-oxaoctane
PubChem CID 78484
IUPAC Name 1-methoxyhexane
SMILES CCCCCCOC

Specifications

Color Colorless
Boiling Point 125°C
Quantity 25 mL
UN Number 3271
Formula Weight 116.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Methyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.