accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 475086-01-2

IUPAC Name: 2-{4-[(5,6-diphenylpyrazin-2-yl)(propan-2-yl)amino]butoxy}-N-methanesulfonylacetamide
Synonyms: selexipag
Molecular Weight (g/mol): 496.63
Molecular Formula: C26H32N4O4S
InChi Key: QXWZQTURMXZVHJ-UHFFFAOYSA-N
SMILES: CC(C)N(CCCCOCC(=O)NS(C)(=O)=O)C1=CN=C(C2=CC=CC=C2)C(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results