accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4759-48-2

IUPAC Name: (2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Synonyms: isotretinoin isotretinoin preparation 15-apo-β-caroten-15-oic acid all-trans-vitamin a acid tretinoina [inn-spanish] retinova acid, trans-retinoic α-acido (argentina) vitamin a acid, trans- tretin M
Molecular Weight (g/mol): 300.44
Molecular Formula: C20H28O2
InChi Key: SHGAZHPCJJPHSC-XFYACQKRSA-N
SMILES: C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C\C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Physical Form 
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (6)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)