accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNONNHNOCH3ClOCH3

CAS RN 476159-98-5

IUPAC Name: N-(2-chloro-5-methoxyphenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
Synonyms: N-(2-chloro-5-methoxyphenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
Molecular Weight (g/mol): 442.94
Molecular Formula: C23H27ClN4O3
InChi Key: WPOXAFXHRJYEIC-UHFFFAOYSA-N
SMILES: COC1=CC(NC2=NC=NC3=CC(OCC4CCN(C)CC4)=C(OC)C=C23)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)