accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 477-75-8

IUPAC Name: pentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene
Synonyms: triptycene
Molecular Weight (g/mol): 254.33
Molecular Formula: C20H14
InChi Key: NGDCLPXRKSWRPY-UHFFFAOYSA-N
SMILES: C1=CC=C2C3C4=CC=CC=C4C(C2=C1)C1=CC=CC=C31
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results