accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4773-13-1

IUPAC Name: bis(propan-2-yl) sulfite
Synonyms: diisopropyl sulfite
Molecular Weight (g/mol): 166.24
Molecular Formula: C6H14O3S
InChi Key: IIKLEMKAKOISSP-UHFFFAOYSA-N
SMILES: CC(C)OS(=O)OC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results