CAS RN 478149-53-0
IUPAC Name:
N-{1-azabicyclo[2.2.2]octan-3-yl}furo[2,3-c]pyridine-5-carboxamide hydrochloride
Synonyms:
N-{1-azabicyclo[2.2.2]octan-3-yl}furo[2,3-c]pyridine-5-carboxamide hydrochloride
Molecular Weight (g/mol):
307.78
Molecular Formula:
C15H18ClN3O2
InChi Key:
YMTPSZUBRSEWNY-UHFFFAOYNA-N
SMILES:
Cl.O=C(NC1CN2CCC1CC2)C1=NC=C2OC=CC2=C1
1
–
1
of
1
results
1
R&D Systems™ PHA 543613 hydrochloride
CAS: 478149-53-0 Molecular Formula: C15H18ClN3O2