CAS RN 479347-85-8
IUPAC Name:
disodium 6-cyano-7-nitroquinoxaline-2,3-bis(olate)
Synonyms:
disodium 6-cyano-7-nitroquinoxaline-2,3-bis(olate)
Molecular Weight (g/mol):
276.12
Molecular Formula:
C9H2N4Na2O4
InChi Key:
YCXDDPGRZKUGDG-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]C1=NC2=CC(C#N)=C(C=C2N=C1[O-])[N+]([O-])=O
1
–
1
of
1
results
1
Tocris Bioscience™ CNQX disodium salt
CAS: 479347-85-8 Molecular Formula: C9H2N4Na2O4