CAS RN 479347-86-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ NBQX disodium salt
Potent AMPA antagonist; more water soluble form of NBQX (Cat. No. 0373)