CAS RN 480-22-8
IUPAC Name:
anthracene-1,8,9-triol
Synonyms:
lasan fna brand of anthralin anthraforte psoradrate cignolin micanol anthralin (substance) stiefel brand 2 of anthralin hamilton brand of anthralin bioglan brand of anthralin
Molecular Weight (g/mol):
226.23
Molecular Formula:
C14H10O3
InChi Key:
YUTJCNNFTOIOGT-UHFFFAOYSA-N
SMILES:
OC1=CC=CC2=CC3=CC=CC(O)=C3C(O)=C12
1
–
1
of
1
results
1
1,8,9-Trihydroxyanthracene 95.0+%, TCI America™
CAS: 480-22-8 Molecular Formula: C14H10O3 Molecular Weight (g/mol): 226.231 MDL Number: MFCD00001250 InChI Key: YUTJCNNFTOIOGT-UHFFFAOYSA-N Synonym: Anthralin, Dithranol, 1,8,9-Anthracenetriol, 1,8-Dihydroxy-9(10H)-anthracenone, DIT PubChem CID: 10187 ChEBI: CHEBI:2756 IUPAC Name: anthracene-1,8,9-triol SMILES: C1=CC2=CC3=C(C(=CC=C3)O)C(=C2C(=C1)O)O