CAS RN 4800-94-6
Carbenicillin (Disodium Salt), Fisher BioReagents
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L Synonym: α-Carboxybenzylpenicillin PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Carbenicillin disodium salt
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L PubChem CID: 20933 ChEBI: CHEBI:34609 SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Thermo Scientific Chemicals Carbenicillin disodium salt, 50 mg/ml in distilled water, sterile-filtered
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Carbenicillin disodium salt, MP Biomedicals™
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L Synonym: α-Carboxybenzylpenicillin PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Thermo Scientific Chemicals Carbenicillin disodium salt, 90%
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Carbenicillin Disodium Salt, MP Biomedicals™
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L Synonym: α-Carboxybenzylpenicillin PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Carbenicillin disodium salt PROMO
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 g/mol MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Carbenicillin Disodium Salt, 5 g, Thermo Scientific Chemicals PROMO
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 g/mol MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Carbenicillin Disodium Salt, 25 g, Thermo Scientific Chemicals PROMO
CAS: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Molecular Weight (g/mol): 422.36 g/mol MDL Number: MFCD00077683 InChI Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L PubChem CID: 20933 ChEBI: CHEBI:34609 IUPAC Name: disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O