Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbenicillin disodium salt PROMO

Catalog No. J6194903F25
Change view
Click to view available options
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
J6194903F25 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J6194903F25 Supplier Thermo Scientific Chemicals Supplier No. J6194903PR/DEL

missing translation for 'validMainframeMsg'

Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities.

General Description

  • Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
  • In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium

Application

  • Due to its antibacterial activity, this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
  • It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4800-94-6
Molecular Formula C17H16N2Na2O6S
Molecular Weight (g/mol) 422.36 g/mol
MDL Number MFCD00077683
InChI Key RTYJTGSCYUUYAL-YCAHSCEMSA-L
PubChem CID 20933
ChEBI CHEBI:34609
IUPAC Name disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Carbenicillin Disodium Salt
Physical Form Powder
Color White
Quantity 1 g
Beilstein 5722128
Merck Index 14,1792
Solubility Information Soluble in Water and Alcohol.
Formula Weight 422.36 amu
Chemical Name or Material Carbenicillin disodium salt
Hazard Category H317-H334-H335
Hazard Statement

GHS H Statement

  • H334-H317
  • May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • May cause an allergic skin reaction.
Precautionary Statement P261-P271-P272-P280g-P285-P302+P352-P304+P340-P333+P313-P342+P311-P363-P501c
EINECSNumber 225-360-8
RTECSNumber ON9105000
TSCA No
Recommended Storage Keep Cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.