accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 480424-80-4

NMgBr

CAS RN 480424-80-4

IUPAC Name: 1-{[2-(bromomagnesio)phenyl]methyl}pyrrolidine
Synonyms: 1-{[2-(bromomagnesio)phenyl]methyl}pyrrolidine
Molecular Weight (g/mol): 264.45
Molecular Formula: C11H14BrMgN
InChi Key: GPUHBRNTFINYSU-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=CC=C1CN1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results