Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, Thermo Scientific™

Catalog No. AC431650500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431650500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431650500 Supplier Thermo Scientific Chemicals Supplier No. 431650500
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480424-80-4 , 109-99-9
Molecular Formula C11H14BrMgN
Molecular Weight (g/mol) 264.45
MDL Number MFCD03840891
InChI Key GPUHBRNTFINYSU-UHFFFAOYSA-M
Synonym 2-1-pyrrolidinylmethyl phenyl magnesium bromide solution, magnesium bromide 2-pyrrolidin-1-yl methyl benzen-1-ide 1/1/1, magnesium 2+ ion 2-pyrrolidin-1-ylmethyl benzen-1-ide bromide, grignard reagent
PubChem CID 16217257
IUPAC Name 1-{[2-(bromomagnesio)phenyl]methyl}pyrrolidine
SMILES Br[Mg]C1=CC=CC=C1CN1CCCC1

Specifications

Color Brown
CAS Min % 4.32, 89.93
CAS Max % 10.07, 95.68
Density 0.9190g/mL
Quantity 50 mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Specific Gravity 0.919
Formula Weight 264.44
Concentration or Composition (by Analyte or Components) 0.15 to 0.35M
Chemical Name or Material [2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.