CAS RN 48209-28-5
CAS RN 48209-28-5
IUPAC Name:
bis(λ¹-copper(1+)) toluene ditrifluoromethanesulfonate
Synonyms:
bis(λ¹-copper(1+)) toluene ditriflate
Molecular Weight (g/mol):
517.36
Molecular Formula:
C9H8Cu2F6O6S2
InChi Key:
ATUUSMBCJWYMEG-UHFFFAOYSA-L
SMILES:
[Cu+].[Cu+].CC1=CC=CC=C1.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Chemicals Copper(I) trifluoromethanesulfonate toluene complex (2:1), 98%
CAS: 48209-28-5 Molecular Formula: C9H8Cu2F6O6S2 Molecular Weight (g/mol): 517.359 MDL Number: MFCD01863766 InChI Key: ATUUSMBCJWYMEG-UHFFFAOYSA-L Synonym: copper i trifluoromethanesulfonate toluene complex,cuotf-toluene,copper i triflate toluene complex,cuprous trifluoromethanesulfonate toluene complex,trifluoromethanesulfonic acid copper i salt toluene complex,copper i trifluoromethanesulfonate toluene complex 2:1 PubChem CID: 11656500 IUPAC Name: copper(1+);toluene;trifluoromethanesulfonate SMILES: CC1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Cu+].[Cu+]