Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) trifluoromethanesulfonate toluene complex (2:1), 98%

Catalog No. AAL1965606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1965606 5 g
AAL1965603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1965606 Supplier Thermo Scientific Chemicals Supplier No. L1965606
  • $432.00
    $367.65 / Each of 1

    Save $64.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 48209-28-5 | C9H8Cu2F6O6S2 | 517.359 g/mol

Copper(I) trifluoromethanesulfonate toluene complex serves as a vital coupling reagent for photo reactions. It is also used in the preparation of polycyclic furans and butyrolactones. Further, it is used as a reagent for the preparation of beta-lactam from beta-amino thiol-esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(I) trifluoromethanesulfonate toluene complex serves as a vital coupling reagent for photo reactions. It is also used in the preparation of polycyclic furans and butyrolactones. Further, it is used as a reagent for the preparation of beta-lactam from beta-amino thiol-esters.

Solubility
Soluble in toluene.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48209-28-5
Molecular Formula C9H8Cu2F6O6S2
Molecular Weight (g/mol) 517.359
MDL Number MFCD01863766
InChI Key ATUUSMBCJWYMEG-UHFFFAOYSA-L
Synonym copper i trifluoromethanesulfonate toluene complex, cuotf-toluene, copper i triflate toluene complex, cuprous trifluoromethanesulfonate toluene complex, trifluoromethanesulfonic acid copper i salt toluene complex, copper i trifluoromethanesulfonate toluene complex 2:1
PubChem CID 11656500
IUPAC Name copper(1+);toluene;trifluoromethanesulfonate
SMILES CC1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Cu+].[Cu+]

Specifications

Melting Point 180°C (decomposition)
Flash Point 76°C (168°F)
Quantity 5 g
UN Number UN1325
Sensitivity Air and moisture sensitive
Solubility Information Soluble in toluene.
Formula Weight 517.37
Percent Purity 98%
Chemical Name or Material Copper(I) trifluoromethanesulfonate toluene complex (2:1)
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.