CAS RN 4837-01-8
CAS RN 4837-01-8
IUPAC Name:
[(2-bromoethyl)sulfanyl]benzene
Synonyms:
[(2-bromoethyl)sulfanyl]benzene
Molecular Weight (g/mol):
217.12
Molecular Formula:
C8H9BrS
InChi Key:
UEFBOQYLXLEJSM-UHFFFAOYSA-N
SMILES:
BrCCSC1=CC=CC=C1
1
–
1
of
1
results
1
2-Bromoethyl Phenyl Sulfide 97.0+%, TCI America™
CAS: 4837-01-8 Molecular Formula: C8H9BrS Molecular Weight (g/mol): 217.12 MDL Number: MFCD00040826 InChI Key: UEFBOQYLXLEJSM-UHFFFAOYSA-N Synonym: 2-bromoethyl phenyl sulfide,2-bromoethyl thio benzene,2-bromoethyl phenyl sulfane,2-bromoethyl sulfanyl benzene,2-bromoethylthiobenzene,benzene, 2-bromoethyl thio,2-bromo-1-phenylthioethane,acmc-1aoyt,2-phenylthioethyl bromide PubChem CID: 78553 IUPAC Name: [(2-bromoethyl)sulfanyl]benzene SMILES: BrCCSC1=CC=CC=C1