accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4837-90-5

IUPAC Name: 4-methoxy-1H-indole
Synonyms: 4-methoxy-1H-indole
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: LUNOXNMCFPFPMO-UHFFFAOYSA-N
SMILES: COC1=C2C=CNC2=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)