Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyindole, 99%, Thermo Scientific Chemicals

Catalog No. AAL1906903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1906903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1906903 Supplier Thermo Scientific Chemicals Supplier No. L1906903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4837-90-5 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4837-90-5
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00009737
InChI Key LUNOXNMCFPFPMO-UHFFFAOYSA-N
Synonym 4-methoxyindole, 1h-indole, 4-methoxy, 4-methoxylindole, 1h-indol-4-yl methyl ether, indole, 4-methoxy, 4-methoxy-indole, zlchem 710, pubchem7433, acmc-1aoxe
PubChem CID 138363
IUPAC Name 4-methoxy-1H-indole
SMILES COC1=CC=CC2=C1C=CN2

Specifications

Melting Point 69°C to 71°C
Boiling Point 181°C to 183°C (24 mmHg)
Quantity 1 g
Beilstein 122456
Sensitivity Light sensitive
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 4-Methoxyindole
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.