CAS RN 4847-93-2
IUPAC Name:
3-(piperidin-1-yl)propane-1,2-diol
Synonyms:
3-(piperidin-1-yl)propane-1,2-diol
Molecular Weight (g/mol):
159.23
Molecular Formula:
C8H17NO2
InChi Key:
MECNWXGGNCJFQJ-UHFFFAOYNA-N
SMILES:
OCC(O)CN1CCCCC1
1
–
1
of
1
results
1
3-Piperidino-1,2-propanediol 96%, Thermo Scientific™
CAS: 4847-93-2 Molecular Formula: C8H17NO2 Molecular Weight (g/mol): 159.229 InChI Key: MECNWXGGNCJFQJ-UHFFFAOYSA-N Synonym: 3-piperidino-1,2-propanediol,3-piperidin-1-yl propane-1,2-diol,1-n-piperidino-2,3-propanediol,3-piperidinopropane-1,2-diol,3-piperidylpropane-1,2-diol,1-piperidino-2,3-propanediol,1,2-propanediol,3-1-piperidinyl,1,2-propanediol, 3-1-piperidinyl,3-1-piperidinyl-1,2-propanediol # PubChem CID: 97824 IUPAC Name: 3-piperidin-1-ylpropane-1,2-diol SMILES: C1CCN(CC1)CC(CO)O