accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4857-44-7

IUPAC Name: calcium bis(2-hydroxy-4-(methylsulfanyl)butanoate)
Synonyms: calcium bis(2-hydroxy-4-(methylsulfanyl)butanoate)
Molecular Weight (g/mol): 338.45
Molecular Formula: C10H18CaO6S2
InChi Key: ABRVDWASZFDIEH-UHFFFAOYNA-L
SMILES: [Ca++].CSCCC(O)C([O-])=O.CSCCC(O)C([O-])=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)