CAS RN 487051-12-7
IUPAC Name:
N-[3-(1-{[4-(3,4-difluorophenoxy)phenyl]methyl}piperidin-4-yl)-4-methylphenyl]-2-methylpropanamide hydrochloride
Synonyms:
N-[3-(1-{[4-(3,4-difluorophenoxy)phenyl]methyl}piperidin-4-yl)-4-methylphenyl]-2-methylpropanamide hydrochloride
Molecular Weight (g/mol):
515.04
Molecular Formula:
C29H33ClF2N2O2
InChi Key:
DEDUDFNRQKUBRH-UHFFFAOYSA-N
SMILES:
Cl.CC(C)C(=O)NC1=CC(C2CCN(CC3=CC=C(OC4=CC(F)=C(F)C=C4)C=C3)CC2)=C(C)C=C1
1
–
1
of
1
results
1
R&D Systems™ SNAP 94847 hydrochloride
CAS: 487051-12-7 Molecular Formula: C29H33ClF2N2O2