accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)(R)OH(S)OHHO(S)HO

CAS RN 488-73-3

IUPAC Name: (1R,2S,4S,5R)-cyclohexane-1,2,3,4,5-pentol
Synonyms: quercitol 2-deoxy-D-chiro-inositol 5-deoxyinositol d-quercitol
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: IMPKVMRTXBRHRB-MBMOQRBOSA-N
SMILES: O[C@@H]1C[C@@H](O)[C@H](O)C(O)[C@H]1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results