CAS RN 4917-76-4
IUPAC Name:
2-[(1,2-dicarboxyethyl)sulfanyl]butanedioic acid
Synonyms:
2-[(1,2-dicarboxyethyl)sulfanyl]butanedioic acid
Molecular Weight (g/mol):
266.22
Molecular Formula:
C8H10O8S
InChi Key:
SASYRHXVHLPMQD-UHFFFAOYNA-N
SMILES:
OC(=O)CC(SC(CC(O)=O)C(O)=O)C(O)=O