accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 491871-58-0

IUPAC Name: 6-(5-bromo-2-hydroxyphenyl)-2-oxo-4-phenyl-1,2-dihydropyridine-3-carbonitrile
Synonyms: 6-(5-bromo-2-hydroxyphenyl)-2-oxo-4-phenyl-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 367.2
Molecular Formula: C18H11BrN2O2
InChi Key: SVSYJTYGPLVUOZ-UHFFFAOYSA-N
SMILES: OC1=C(C=C(Br)C=C1)C1=CC(C2=CC=CC=C2)=C(C#N)C(=O)N1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results