accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)(R)N(S)(R)

CAS RN 492-08-0

IUPAC Name: (1R,2R,9R,10S)-7,15-diazatetracyclo[7.7.1.0²,⁷.0¹⁰,¹⁵]heptadecane
Synonyms: L-sparteine
Molecular Weight (g/mol): 234.39
Molecular Formula: C15H26N2
InChi Key: SLRCCWJSBJZJBV-TUVASFSCSA-N
SMILES: C1CCN2C[C@H]3C[C@H](CN4CCCC[C@H]34)[C@@H]2C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)