CAS RN 4926-90-3
IUPAC Name:
1-ethyl-1-methylcyclohexane
Synonyms:
cyclohexane, 1-ethyl-1-methyl-
Molecular Weight (g/mol):
126.24
Molecular Formula:
C9H18
InChi Key:
YPJRYQGOKHKNKZ-UHFFFAOYSA-N
SMILES:
CCC1(C)CCCCC1
1
–
1
of
1
results
1
1-Ethyl-1-Methylcyclohexane, SPEX CertiPrep™
CAS: 4926-90-3 Molecular Formula: C9H18 Molecular Weight (g/mol): 126.243 InChI Key: YPJRYQGOKHKNKZ-UHFFFAOYSA-N PubChem CID: 35411 IUPAC Name: 1-ethyl-1-methylcyclohexane SMILES: CCC1(CCCCC1)C