CAS RN 493-52-7
Thermo Scientific Chemicals Methyl Red, ACS reagent
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2,2-[4-(Dimethylamino)phenylazo]benzoic acid,C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Thermo Scientific Chemicals Methyl Red, pure, indicator
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2,2-[4-(Dimethylamino)phenylazo]benzoic acid,C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red, Neutral, Reagent, ACS, Spectrum™ Chemical
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.30 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N IUPAC Name: 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
Methyl red, For ACS analysis, MP Biomedicals™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: 2-(4-Dimethylaminophenylazo)benzoic acid,4-Dimethylaminoazobenzene-2'-carboxylic acid PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Thermo Scientific Chemicals Methyl Red
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Thermo Scientific Chemicals Methyl Red, ACS
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: C.I. 13020 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red (0.1% in ca. 95% Ethanol) [for Titration], TCI America™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: methyl red,o-methyl red,c.i. acid red 2,acid red 2,cerven methylova,cerven kysela 2,4-dimethylamino-2'-carboxylazobenzene,2-carboxy-4'-dimethylamino azobenzene,2-4-dimethylaminophenylazo benzoic acid,ci acid red 2 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red (0.04% in Water) [for pH Determination], TCI America™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: methyl red,o-methyl red,c.i. acid red 2,acid red 2,cerven methylova,cerven kysela 2,4-dimethylamino-2'-carboxylazobenzene,2-carboxy-4'-dimethylamino azobenzene,2-4-dimethylaminophenylazo benzoic acid,ci acid red 2 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red 98.0+%, TCI America™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: methyl red,o-methyl red,c.i. acid red 2,acid red 2,cerven methylova,cerven kysela 2,4-dimethylamino-2'-carboxylazobenzene,2-carboxy-4'-dimethylamino azobenzene,2-4-dimethylaminophenylazo benzoic acid,ci acid red 2 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red 92.0+%, TCI America™
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: methyl red,o-methyl red,c.i. acid red 2,acid red 2,cerven methylova,cerven kysela 2,4-dimethylamino-2'-carboxylazobenzene,2-carboxy-4'-dimethylamino azobenzene,2-4-dimethylaminophenylazo benzoic acid,ci acid red 2 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble), Reagents
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.30 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid IUPAC Name: 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
Methyl Red Indicator, Reag. Ph. Eur., Solstice
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red Solution, Solstice
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Methyl Red, Free Acid (Certified), Fisher Chemical
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.30 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2,o-(p-Dimethylaminophenylazo)benzoic Acid PubChem CID: 10303 IUPAC Name: 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O