Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methyl Red, pure, indicator

Catalog No. AC264445000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264445000 500 g Glass Bottle
AC264440250 25 g Glass Bottle
AC264441000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC264445000 Supplier Thermo Scientific Chemicals Supplier No. 264445000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Methyl Red, C15H15N3O2, CAS Number-493-52-7, Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020, 25g, 269.3, Pure, Warning, GHS H Statement: Harmful if swallowed. | CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 178.0°C to 182.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Extinction 650 min. (430nm, in buffer)
Linear Formula (CH3)2NC6H4N=NC6H4CO2H
Visual Transition Interval from pH 4.2 (red) to pH 6.2 (yellow)
Color Red
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol, acetic acid
Quantity 500 g
Physical Form Powder
Formula Weight 269.3
Grade Pure
Chemical Name or Material Methyl Red
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.