Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red, Free Acid (Certified), Fisher Chemical

Catalog No. M29610 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M296-10 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M296-10

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2, o-(p-Dimethylaminophenylazo)benzoic Acid
PubChem CID 10303
IUPAC Name 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O

Specifications

Melting Point 178°C
Linear Formula 4-(CH3)2N-C6H4-N:N-2-(CO2H)C6H4
Identification Pass Test
Formula Weight 269.3
Grade Certified
Packaging Glass Bottle
Visual Transition Interval Pass Test
Color Index No. 13020
Color Violet
Quantity 10 g
Physical Form Solid
Chemical Name or Material Methyl Red, Free Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.