accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3O

CAS RN 4944-60-9

IUPAC Name: 9-(acetyloxy)nonyl acetate
Synonyms: nonanediol, diacetate
Molecular Weight (g/mol): 244.33
Molecular Formula: C13H24O4
InChi Key: JAEVCKMMGIFOCL-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCCCCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results