accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49539-88-0

IUPAC Name: trimethoxy(2-phenylethyl)silane
Synonyms: trimethoxy(2-phenylethyl)silane
Molecular Weight (g/mol): 226.35
Molecular Formula: C11H18O3Si
InChi Key: UBMUZYGBAGFCDF-UHFFFAOYSA-N
SMILES: CO[Si](CCC1=CC=CC=C1)(OC)OC
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)