Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoxy(2-phenylethyl)silane 73.0+%, TCI America™
SDP

Catalog No. T286925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T286925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T286925G Supplier TCI America Supplier No. T286925G

Chemical Identifiers

CAS 49539-88-0
Molecular Formula C11H18O3Si
Molecular Weight (g/mol) 226.347
MDL Number MFCD00053852
InChI Key UBMUZYGBAGFCDF-UHFFFAOYSA-N
Synonym Trimethoxy(phenethyl)silane
PubChem CID 170789
IUPAC Name trimethoxy(2-phenylethyl)silane
SMILES CO[Si](CCC1=CC=CC=C1)(OC)OC

Specifications

Boiling Point 122°C
Quantity 25 g
Formula Weight 226.35
Percent Purity ≥73.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(2-phenylethyl)silane [contains ca. 25% Trimethoxy(1-phenylethyl)silane]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.