CAS RN 49553-93-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ PEI STAR-Go™ transfection reagent
Aqueous solution of polyethylenimine (PEI) transfection reagent
2
Thermo Scientific™ Polyethyleneimine hydrochloride, linear, M.W. 40,000
CAS: 49553-93-7 | -HN-[CH{2}CH{2}NH-]+n+.HCl
3
Tocris Bioscience™ PEI STAR™ transfection reagent
Polyethylenimine (PEI) transfection reagent, chemically-defined