accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHH2NSN

CAS RN 49600-34-2

IUPAC Name: (6-methylpyridin-2-yl)thiourea
Synonyms: 6-methylpyridin-2-ylthiourea
Molecular Weight (g/mol): 167.23
Molecular Formula: C7H9N3S
InChi Key: LEWSYNYKTYXRHP-UHFFFAOYSA-N
SMILES: CC1=CC=CC(NC(N)=S)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results