accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49783-72-4

IUPAC Name: 3-methyl-1,2-oxazole-5-carbonyl chloride
Synonyms: 3-methyl-1,2-oxazole-5-carbonyl chloride
Molecular Weight (g/mol): 145.54
Molecular Formula: C5H4ClNO2
InChi Key: VUBCNGCMOPTDED-UHFFFAOYSA-N
SMILES: CC1=NOC(=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results